missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine, TRC

Product Code. 30381189
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381189 5 g -
30373303 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381189 Supplier Toronto Research Chemicals Supplier No. TRCD4789405G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-39-4
Molecular Formula C12 H11 N
Molecular Weight (g/mol) 169.22
Synonym Benzenamine, N-phenyl-, Diphenylamine (8CI), N-Phenylbenzenamine, Anilinobenzene, Antioxidant DPA, Benzene, (phenylamino)-, DBA, DFA, DPA, N,N-Diphenylamine, N-Phenylaniline, NSC 215210, Naugalube 428L, No-Scald
IUPAC Name N-phenylaniline
SMILES N(c1ccccc1)c2ccccc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H11N/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10,13H
Formula Weight 169.0891
Chemical Name or Material Diphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.