missing translation for 'onlineSavingsMsg'
Learn More

Diphenylborinic anhydride, 95%

Product Code. 15451788
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15451788 1 g 1g
15471788 250 mg 250mg
15461788 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451788 Supplier Thermo Scientific Alfa Aesar Supplier No. H55509.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4426-21-5 | C24H20B2O | 346.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4426-21-5
Molecular Formula C24H20B2O
Molecular Weight (g/mol) 346.04
MDL Number MFCD00042663
InChI Key SNQFEECGHGUHBK-UHFFFAOYSA-N
Synonym diphenylborinic anhydride, oxybis diphenylborane, diphenylboranyloxy diphenyl borane, diphenylboronic anhydride, 1,1,3,3-tetraphenyldiboroxane, diphenylboranyl diphenylborinate, tetraphenyldiboroxane, borane, oxybis diphenyl, diphenyl borinic anhydride, d03twr
PubChem CID 596810
IUPAC Name diphenylboranyloxy(diphenyl)borane
SMILES O(B(C1=CC=CC=C1)C1=CC=CC=C1)B(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 135°C to 140°C
Color White to Yellow
Linear Formula (C6H5)2BOB(C6H5)2
Quantity 1 g
Beilstein 2950331
Sensitivity Moisture sensitive
Formula Weight 346.04
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Diphenylborinic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.