missing translation for 'onlineSavingsMsg'
Learn More

Diphenyleneiodonium chloride, 97+%

Product Code. 10633352 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10633352 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10633352 Supplier Thermo Scientific Acros Supplier No. 328660250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4673-26-1 | C12H8ClI | 314.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4673-26-1
Melting Point 312.0°C to 322.0°C
Color White
Assay Percent Range 97+%
Molecular Formula C12H8ClI
MDL Number MFCD00214165
Quantity 25 mg
Synonym diphenyleneiodonium chloride, dibenziodolium chloride, unii-7m9d81yz2n, dpi, 8? 3-iodatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2,4,6,9,11-hexaen-8-ylium chloride, dibenziodolium, chloride, d0n0gj, c12h8i.cl, dibenzo b,d iodolium chloride, 2,2'-biphenylyleneiodonium chloride
Solubility Information Solubility in water: .. Other solubilities: soluble in dmso
InChI Key FCFZKAVCDNTYID-UHFFFAOYSA-M
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3[I+]2.[Cl-]
Molecular Weight (g/mol) 314.55
PubChem CID 2733504
ChEBI CHEBI:77967
Formula Weight 314.55
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material Diphenyleneiodonium chloride
Show More Show Less
compliance-icons
Product Identifier
  • Diphenyleneiodonium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.