missing translation for 'onlineSavingsMsg'
Learn More

Diphenylmethylphosphine, polymer-supported, 0.9-1.4 mmol/g on polystyrene

Product Code. 15430857
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430857 25 g 25g
15410857 1 g 1g
15420857 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15430857 Supplier Thermo Scientific Alfa Aesar Supplier No. L19477.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1486-28-8 | C13H13P | 200.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1486-28-8
Molecular Formula C13H13P
Molecular Weight (g/mol) 200.22
MDL Number MFCD00008508
InChI Key UJNZOIKQAUQOCN-UHFFFAOYSA-N
Synonym methyldiphenylphosphine, diphenylmethylphosphine, methyl diphenyl phosphine, phosphine, methyldiphenyl, methyldiphenylphsphine, diphenylphosphinomethane, methyldiphenylphoshine, pubchem6421, methyl diphenylphosphine, methyldi-phenylphosphine
PubChem CID 73879
IUPAC Name methyl(diphenyl)phosphane
SMILES CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 25 g
Chemical Name or Material Diphenylmethylphosphine, polymer-supported, 0.9 to 1.4mmol/g on polystyrene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.