missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphine Oxide, TRC

Product Code. 30379318
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379318 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379318 Supplier Toronto Research Chemicals Supplier No. TRCD49274850MG
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylphosphine Oxide
CAS 4559-70-0
Molecular Formula C12H11OP
Quantity 50 mg
Synonym Diphenylphosphine Oxide, Diphenylphosphine oxide (ACI), (Phenylphosphoroso)benzene, DPPO, NSC 77604, OX 2, OX 2 (flame retardant)
InChI Formula InChI=1S/C12H11OP/c13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,14H
SMILES O=P(c1ccccc1)c2ccccc2
IUPAC Name phenylphosphonoylbenzene
Molecular Weight (g/mol) 202.18
Formula Weight 202.0548
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.