missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphorazidate, TRC

Product Code. 30385648
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385648 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385648 Supplier Toronto Research Chemicals Supplier No. TRCD49164010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylphosphorazidate
CAS 26386-88-9
Molecular Formula C12H10N3O3P
Quantity 10 g
Synonym Azidodiphenoxyoxophosphorane, DPPA, Diphenyl Azidophosphate, Diphenyl Phosphorazide, Diphenylphosphoryl Azide, O,O-Diphenylphosphoryl Azide, Phosphoric Acid Diphenyl Ester Azide
InChI Formula InChI=1S/C12H10N3O3P/c13-14-15-19(16,17-11-7-3-1-4-8-11)18-12-9-5-2-6-10-12/h1-10H
SMILES [N-]=[N+]=NP(=O)(Oc1ccccc1)Oc2ccccc2
Molecular Weight (g/mol) 275.2
Formula Weight 275.046
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.