missing translation for 'onlineSavingsMsg'
Learn More

Diphenylsilane, 97%

Product Code. 10105960 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10105960 5 g Glass bottle 5g
10032001 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10105960 Supplier Thermo Scientific Acros Supplier No. 329770050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 775-12-2 | C12H12Si | 184.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 775-12-2
Molecular Formula C12H12Si
Molecular Weight (g/mol) 184.31
MDL Number MFCD00003002
InChI Key BPYFPNZHLXDIGA-UHFFFAOYSA-N
Synonym diphenylsilane, silane, diphenyl, diphenylsilicon, benzene, 1,1'-silylenebis, diphenyl silane, di phenyl silicon, diphenylsilylene radical, ph 2sih2
PubChem CID 6327659
IUPAC Name cyclohexa-2,5-dien-1-ylidene(phenyl)silanide
SMILES C1=CC=C(C=C1)[Si-]=C2C=C[CH+]C=C2

Specifications

Color Colorless
Density 0.9900g/mL
Boiling Point 95°C to 97°C (13.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C6H5)2SiH2
Refractive Index 1.5780 to 1.58
Quantity 5 g
Fieser 01,348; 13,153; 15,151; 16,160; 17,132
Specific Gravity 0.99
Solubility Information Solubility in water: insoluble in water
Formula Weight 184.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diphenylsilane, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Diphenylsilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.