missing translation for 'onlineSavingsMsg'
Learn More

Diphenyltin dichloride, 90+%

Product Code. 11314829
missing translation for 'orderingAttributeHoverText'
Quantity:
2 g
10 g
50 g
missing translation for 'unitSize'
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11314829

missing translation for 'mfr': Thermo Scientific Alfa Aesar 071139.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1135-99-5 | C12H12Cl2Sn | 345.84 g/mol

Used in the manufacture of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1135-99-5
Molecular Formula C12H12Cl2Sn
Molecular Weight (g/mol) 345.84
MDL Number MFCD00000516
InChI Key WSXPROXKZIMZIU-UHFFFAOYSA-N
Synonym diphenyltin dichloride, stannane, dichlorodiphenyl, dichlorodiphenyltin, diphenyldichlorotin, diphenyltin chloride, diphenylstannyl dichloride, tin, diphenyl-, dichloride, dichloro diphenyl stannane, diphenylstannium dichloride, unii-0y7mz4k0dr
PubChem CID 14342
IUPAC Name dichloro(diphenyl)stannane
SMILES Cl.Cl.[SnH2](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 41°C to 43°C
Color White
Boiling Point 333°C to 337°C (decomposition)
Flash Point >150°C (302°F)
Linear Formula (C6H5)2SnCl2
Quantity 10 g
Sensitivity Moisture sensitive
Solubility Information Sparingly Soluble (2.4E-3g/L) (25°C),Calc.
Formula Weight 343.81
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Diphenyltin dichloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.