missing translation for 'onlineSavingsMsg'
Learn More

Diphylline, MedChemExpress

Product Code. 16488116
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488116 100 mg 100mg
16442036 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488116 Supplier Medchem Express Supplier No. HYB0128
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Diphylline (Diprophylline) is a potent A1/A2 adenosine receptor antagonist and cyclic nucleotide phosphodiesterase inhibitor. Diphylline, a xanthine derivative, is a bronchodilator and vasodilator drug and has the potential for chronic bronchitis and emphysema.

Diphylline (Diprophylline) is a potent A1/A2 adenosine receptor antagonist and cyclic nucleotide phosphodiesterase inhibitor. Diphylline, a xanthine derivative, is a bronchodilator and vasodilator drug and has the potential for chronic bronchitis and emphysema.

Specifications

Chemical Name or Material Diphylline
CAS 479-18-5
Color White
Purity Grade Notes Research
Molecular Formula C10H14N4O4
Quantity 100 mg
Synonym Diprophylline
Solubility Information DMSO : ≥ 50 mg/mL (196.66 mM) ∣H2O : 20 mg/mL (78.67 mM; Need ultrasonic)
SMILES O=C(N1C)N(C)C2=C(N(CC(O)CO)C=N2)C1=O
Molecular Weight (g/mol) 254.24
Formula Weight 254.24
Percent Purity 99.07%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.