missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene glycol, 99%, mixture of isomers

Product Code. 10289550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
3 kg
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1kg
3kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10289550 1 kg Glass Bottle 1kg
10742212 3 kg Plastic drum 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10289550 Supplier Thermo Scientific Acros Supplier No. 409150010
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25265-71-8 | C6H14O3 | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Note: The chemical structure above shows only one of the molecules included in this mixture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25265-71-8
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00051023

Specifications

Color Orange-Red to Yellow
Density 1.0200g/mL
Boiling Point 229.0°C to 232.0°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH2OCH2CH(OH)CH3
Refractive Index 1.4400 to 1.4420
Quantity 1 kg
Specific Gravity 1.02
Solubility Information Solubility in water: miscible. Other solubilities: soluble in ethanol
Formula Weight 134.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dipropylene glycol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.