missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene glycol, mixture of isomers, 99%

Product Code. 11402998
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11402998 2500 g 2500g
11492988 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11402998 Supplier Thermo Scientific Alfa Aesar Supplier No. A18716.0E
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25265-71-8 | C6H14O3 | 134.18 g/mol

Dipropylene glycol, mixture of isomers is used as a solvent, plasticizer, polymerization initiator and monomer. It acts as an additive used in perfumes, skin and hair care products. It is an active ingredient in commercial fog fluid, which is used as an entertainment in fog machines. Further, it is used as an intermediate in polyurethane polyols, alkyd resins and non-reinforced polyesters. In addition to this, it is used in dyes, paints, hydraulic brake fluids and cutting oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dipropylene glycol, mixture of isomers is used as a solvent, plasticizer, polymerization initiator and monomer. It acts as an additive used in perfumes, skin and hair care products. It is an active ingredient in commercial fog fluid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25265-71-8

Specifications

Melting Point -32°C
Density 1.022
Boiling Point 231°C to 235°C
Odor Odorless
Molecular Formula HOC3H6OC3H6OH
Refractive Index 1.441
MDL Number MFCD00051023
Quantity 2500 g
Synonym Bis(hydroxypropyl) ether; Oxybispropanol
Solubility Information Miscible with cold water,hot water,methanol,diethyl ether and ethanol.
Formula Weight 134.18
Percent Purity 99%
Chemical Name or Material Dipropylene glycol, mixture of isomers
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.