missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene Glycol (Mixture of Isomers), TRC

Product Code. 30388307
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388307 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388307 Supplier Toronto Research Chemicals Supplier No. TRCD492755100G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dipropyleneglycol
CAS 25265-71-8
Molecular Formula ((C6 H14 O3)c(C6 H14 O3)c(C6 H14 O3)c)mix
Quantity 100 g
Synonym Oxybispropanol, Dipropylene glycol, ADK DPG-RF, DPG-FC, DPG-RF, Dowanol DPG, Dipropyleneglycol
InChI Formula InChI=1S/C6H14O3/c1-3-5(7)9-6(8)4-2/h5-8H,3-4H2,1-2H3
SMILES CC(O)COCC(C)O
Molecular Weight (g/mol) 402.52
Formula Weight 402.2829
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.