missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene Glycol Monomethyl Ether (Mixture of Isomers), TRC

Product Code. 30384889
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384889 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384889 Supplier Toronto Research Chemicals Supplier No. TRCD495808500MG
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dipropylene glycol-monoethyl ether
CAS 34590-94-8
Molecular Formula C7 H16 O3
Quantity 500 mg
Synonym Propanol, 1(or 2)-(2-methoxymethylethoxy)-, Dipropylene glycol monomethyl ether (6CI,7CI), 1(or 2)-(2-Methoxymethylethoxy)propanol, Arcosolv DPM, DPM, Dipropylene glycol methyl ether, Dowanol DPM, Forguard M, Glysolv DPM, Hisolve DPM, Kino-red, Methoxypropoxypropanol, Methyl Dipropasol, Methyl Propylene Di Glycol, Oxybispropanol methyl ether, PPG-2 methyl ether
InChI Formula InChI=1S/C7H16O3/c1-9-5-3-7-10-6-2-4-8/h8H,2-7H2,1H3
SMILES CCCCOCC(C)OCC(C)O
IUPAC Name 3-(3-methoxypropoxy)propan-1-ol
Molecular Weight (g/mol) 148.2
Formula Weight 148.1099
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.