missing translation for 'onlineSavingsMsg'
Learn More

Diquafosol tetrasodium, MedChemExpress

Product Code. 16488548
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488548 10 mg 10mg
16448386 100 mg 100mg
16434417 50 mg 50mg
16411028 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488548 Supplier Medchem Express Supplier No. HYB0606
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Diquafosol tetrasodium is a P2Y2 receptor agonist that stimulates fluid and mucin secretion on the ocular surface, as a topical treatment of dry eye disease.

Diquafosol tetrasodium is a P2Y2 receptor agonist that stimulates fluid and mucin secretion on the ocular surface, as a topical treatment of dry eye disease.

Specifications

Chemical Name or Material Diquafosol tetrasodium
CAS 211427-08-6
Color White
Purity Grade Notes Research
Molecular Formula C18H22N4Na4O23P4
Quantity 10 mg
Synonym INS365
Solubility Information H2O : 100 mg/mL (113.87 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.14 mM; Need ultrasonic)
SMILES O[C@H]1[C@H](N(C=CC2=O)C(N2)=O)O[C@H](COP(OP(OP(OP(OC[C@H]3O[C@@H](N(C=CC4=O)C(N4)=O)[C@H](O)[C@@H]3O)(O[Na])=O)(O[Na])=O)(O[Na])=O)(O[Na])=O)[C@H]1O
Molecular Weight (g/mol) 878.23
Formula Weight 878.23
Percent Purity 98.49%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.