missing translation for 'onlineSavingsMsg'
Learn More

Diroximel Fumarate, TRC

Product Code. 30133306
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133306 100 mg 100mg
30138294 500 mg 500mg
30009544 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133306 Supplier Toronto Research Chemicals Supplier No. TRCD491485100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1577222-14-0
Molecular Formula C11 H13 N O6
Molecular Weight (g/mol) 255.22
Synonym 2-(2,5-Dioxopyrrolidin-1-yl)ethyl Methyl Fumarate
IUPAC Name O4-[2-(2,5-dioxopyrrolidin-1-yl)ethyl] O1-methyl (E)-but-2-enedioate
SMILES COC(=O)\C=C\C(=O)OCCN1C(=O)CCC1=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H13NO6/c1-17-10(15)4-5-11(16)18-7-6-12-8(13)2-3-9(12)14/h4-5H,2-3,6-7H2,1H3/b5-4+
Formula Weight 255.0743
Chemical Name or Material Diroximel fumarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.