missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Discofix™ 3-Way Stopcock

Product Code. 17534546
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17534546 Blue Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17534546 Supplier B Braun™ Supplier No. 4095111
  • ONLINE EXCLUSIVE PRICE
    48.11 €
    43.23 € / Pack of 50
    Save 4.88 €
    10% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lock swivels both axially and radially for secure and speedy connection. B Braun™ Discofix™ 3-Way Stopcock is suitable for all current pressure infusion systems.

Stopcock system for infusion therapy and monitoring
  • Suitable for all current pressure infusion systems
  • Continuous straight-flow channels for accurate pressure monitoring
  • Swivel lock moves both axially and radially for secure and speedy connection

Specifications

Color Blue
For Use With (Application) Infusion therapy and monitoring
Product Type Valve
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.