Learn More
Ibidi μ-Dish 35 mm, Low ibiTreat
Click to view available options
Unit Size:
Pack of 60
Description
- A 35 mm imaging dish with low walls
- used in high-end microscopy and micromanipulation
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Specifications
Specifications
| Gridded | Not Gridded |
| Sterility | Sterile |
| Type | Cell Culture Dish |
| Quantity | 60 |
| Diameter (Metric) | 35 mm |