missing translation for 'onlineSavingsMsg'
Learn More

Day Impex™ Virkon™ Disinfectant Virucidal Tablets

Product Code. 12328667
Click to view available options
Quantity:
10 Tablets/Tub, 20 Tubs/Cs.
50 Tablets/Tub, 12 Tubs/Cs.
Unit Size:
200 tablets
600 tablets
This item is not returnable. View return policy

Product Code. 12328667

Brand: Day Impex™ 330012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Clean and disinfect work surfaces and equipment in a single, one-step operation. Day Impex™ Virkon™ Disinfectant Virucidal Tablets provide the widest proven spectrum of any disinfectant.

Use biocides safely. Always read the label and product information before use.
  • Widest proven spectrum of any disinfectant - proven effective in independent tests against all 18 virus families affecting man and animals, including AIDS (HIV) and Hepatitis B
  • Excellent bactericide and fungicide - cleans and disinfects work surfaces and equipment in a single, one-step operation
  • Can be applied in powder form direct to spillages of blood and other body fluids
  • Easily dissolves to make a liquid disinfectant
  • Odourless, non-irritant to skin and eyes, non-toxic vapour phase, exceptionally low oral toxicity
  • Under COSHH requirements, no maximum exposure limit is required
  • Non-corrosive, non-bleaching and biodegradable
  • Any viral disease transmitted via surfaces or instruments contaminated with infected blood, body fluids, faeces or saliva can be controlled by correct application For cleaning and disinfecting work surfaces and equipment.

Specifications

Product Type Virucidal Disinfectant
Form Tablet
For Use With (Application) For Cleaning and Disinfecting Work Surfaces and Equipment
Includes 5 g/Tablet
Quantity 10 Tablets/Tub, 20 Tubs/Cs.
compliance-icons
Product Identifier
  • Rely+On Virkon Tablets
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.