missing translation for 'onlineSavingsMsg'
Learn More

Disodium Bathocuproine Disulfonate (Mixture of Regioisomers), TRC

Product Code. 30344432
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30344432 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344432 Supplier Toronto Research Chemicals Supplier No. TRCB1187831G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bathocuproine Disulfonic Acid Disodium Salt (Mixture of regioisomers)
CAS 52698-84-7
Molecular Formula C26 H18 N2 O6 S2 . 2Na
Quantity 1 g
Synonym BCS, Bathocupproine Disulfonate Disodium, Bathocuproinesulfonate, Disodium Bathocuproine Disulfonate, Disodium Bathocuproine Disulfonate, (2,9-Dimethyl-1,10-phenanthroline-4,7-diyl)bis-benzenesulfonic Acid Disodium
InChI Formula InChI=1S/C26H20N2O6S2.2Na/c1-15-25(35(29,30)31)21(17-9-5-3-6-10-17)19-13-14-20-22(18-11-7-4-8-12-18)26(36(32,33)34)16(2)28-24(20)23(19)27-15;;/h3-14H,1-2H3,(H,29,30,31)(H,32,33,34);;/q;2*+1/p-2
SMILES [Na+].[Na+].Cc1nc2c(ccc3c(c4ccccc4)c(c(C)nc23)S(=O)(=O)[O-])c(c5ccccc5)c1S(=O)(=O)[O-]
IUPAC Name disodium;2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate
Molecular Weight (g/mol) 564.54
Formula Weight 564.0402
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.