missing translation for 'onlineSavingsMsg'
Learn More

Disodium hydrogen phosphate, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10020400 Shop All J.T.Baker Products
Change view
Click to view available options
:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10020400 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10020400 Supplier JT Baker Supplier No. 0306.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Meets ACS Specifications

Specifications

Melting Point 250°C
CAS 7558-79-4
pH 8.7 to 9.3 (5% Solution at 25°C)
Quantity 1 kg
Assay Percent Range min. 99.0%
Molecular Formula HNa2O4P
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES [Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium hydrogen phosphate
Molecular Weight (g/mol) 141.96
Loss on Drying max. 0.2%
Density 1.53 g/cm³ (20°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.