missing translation for 'onlineSavingsMsg'
Learn More

Disodium Monooctyl Sulfosuccinate, TRC

Product Code. 30134029
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134029 1 g 1g
30133651 500 mg 500mg
30006680 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134029 Supplier Toronto Research Chemicals Supplier No. TRCS6456101G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86878-53-7
Molecular Formula C12H20Na2O7S
Molecular Weight (g/mol) 354.33
Synonym Docusate Sodium Impurity, 1-(2-Ethylhexyl)ester-2-sulfo-butanedioic Acid Sodium Salt;
IUPAC Name disodium;4-(2-ethylhexoxy)-4-oxo-3-sulfonatobutanoate
SMILES O=C(C(S(=O)(O[Na])=O)CC(O[Na])=O)OCC(CCCC)CC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H22O7S.2Na/c1-3-5-6-9(4-2)8-19-12(15)10(7-11(13)14)20(16,17)18;;/h9-10H,3-8H2,1-2H3,(H,13,14)(H,16,17,18);;/q;2*+1/p-2
Formula Weight 354.0725
Percent Purity >95%
Chemical Name or Material Disodium Monooctyl Sulfosuccinate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.