missing translation for 'onlineSavingsMsg'
Learn More

Disodium NADP (~90%), TRC

Product Code. 30471768
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471768 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471768 Supplier Toronto Research Chemicals Supplier No. TRCN242920250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disodium NADP
CAS 24292-60-2
Molecular Formula C21 H26 N7 O17 P3 . 2 Na
Quantity 250 mg
Synonym NAD Sodium Phosphate, Sodium (2R,3R,4R,5R)-2-(6-Amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)(hydroxy)phosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl Phosphate
InChI Formula InChI=1S/C21H28N7O17P3.2Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37,
SMILES [Na+].[Na+].NC(=O)c1ccc[n+](c1)[C@@H]2O[C@H](COP(=O)([O-])OP(=O)([O-])OC[C@H]3O[C@H]([C@H](OP(=O)(O)[O-])[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O
IUPAC Name disodium;[(2R,3R,4R,5R)-2-(6-aminopurin-9-yl)-5-[[[[(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]-4-hydroxyoxolan-3-yl] hydrogen phosphate
Molecular Weight (g/mol) 787.369
Formula Weight 787.039
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.