Learn More
BRAND™ Dispensing Cylinder with Valve Block
Product Code. 11663219 Shop All Brand Products
Click to view available options
For Use With (Equipment):
10 mL Burette
50 mL Burette
Unit Size:
Each
Product Code. 11663219
Brand: BRAND™ 707537
Only null left
Add to basket
Add to basket
For use with Titrette™ Bottletop Burette. Accommodates all serial number ranges.
  • For use with Titrette™ Bottletop Burette
  • Fits all serial number ranges
  • *For 50mL instruments with serial numbers beginning with 01-12 AND letters A-J, must order both Dispensing cylinder and Titrating tube as a pair.
TRUSTED_SUSTAINABILITY
Specifications
Type Dispensing Cylinder
Includes Valve head for titrette
For Use With (Equipment) 50 mL Burette