missing translation for 'onlineSavingsMsg'
Learn More

Disperse Yellow 23 (Technical Grade), TRC

Product Code. 30003235
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003235 50 mg 50mg
30010504 250 mg 250mg
30012095 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003235 Supplier Toronto Research Chemicals Supplier No. TRCD49449850MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6250-23-3
Molecular Formula C18 H14 N4 O
Molecular Weight (g/mol) 302.33
Synonym 4-[2-[4-(2-Phenyldiazenyl)phenyl]diazenyl]phenol, C.I. Disperse Yellow 23, 4-[[4-(Phenylazo)phenyl]azo]phenol, p-[[p-(Phenylazo)phenyl]azo]phenol, Acetoquinone Light Yellow 3RLLZ, Artisil Yellow RGFL, C.I. 26070, Calcophen Yellow 4RL, Celliton Fast Yellow 4RL-CF, Cibacet Yellow 2RG, DTNW 20, Dianix Yellow 5R, Disperse Yellow 23, Disperse Yellow RGFL, Esteroquinone Light Yellow 3RLL, Fantagen Yellow 3RL, Fenacet Fast Yellow 4R, Foron G, Foron Yellow E-RGF 2, Foron Yellow E-RGFL, Foron Yellow RGFL, Foron Yellow RGFL ultra-dispersed, Intrasil Yellow 5R, Latyl Yellow 4RL, NSC 45565, Nyloquinone Yellow 3R, Ostacet Yellow E-L 5R, SRA Fast Golden Yellow XIII, Setacyl Yellow 3RN, Setacyl Yellow P 3RL, Terasil Golden Yellow R, Terasil Yellow 2RG, Yohao Disperse Yellow E-R, p-Hydroxy-p-bis(azobenzene)
IUPAC Name 4-[(E)-[4-[(E)-phenyldiazenyl]phenyl]diazenyl]phenol
SMILES Oc1ccc(cc1)N=Nc2ccc(cc2)N=Nc3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H14N4O/c23-18-12-10-17(11-13-18)22-21-16-8-6-15(7-9-16)20-19-14-4-2-1-3-5-14/h1-13,23H/b20-19+,22-21+
Formula Weight 302.1168
Chemical Name or Material Disperse Yellow 23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.