Learn More
HUBER Displacement Insert
Product Code. 10781664
missing translation for 'orderingAttributeHoverText'
Length (Metric):
85 mm
124 mm
172 mm
194 mm
245 mm
260 mm
275 mm
For Use With (Equipment):
CC-205B
CC-308B
CC-315B
CC-410, CC-410 WL
CC510, CC515, CC905, CC905W, CC906W
Ministat 125/230 W
Ministat 240/240 W
missing translation for 'unitSize'
Each
Product Code. 10781664
missing translation for 'mfr': HUBER 6293
Only null left
Add to basket
Add to basket
For use with refrigeration bath circulators
Displacement insert
Specifications
For Use With (Equipment) CC-410, CC-410 WL
Type Displacement Insert
Length (Metric) 275 mm