Learn More
HUBER Displacement Insert
Product Code. p-8013464
Click to view available options
Length (Metric):
85 mm
124 mm
172 mm
194 mm
245 mm
260 mm
275 mm
For Use With (Equipment):
CC-205B
CC-308B
CC-315B
CC-410, CC-410 WL
CC510, CC515, CC905, CC905W, CC906W
Ministat 125/230 W
Ministat 240/240 W
Unit Size:
Each
Product Code. 10794616
Brand: HUBER 31973
Only null left
Add to basket
Add to basket
For use with refrigeration bath circulators
Displacement insert
Specifications
For Use With (Equipment) CC-308B
Type Displacement Insert
Length (Metric) 124 mm