missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Disposable Centrifuge Tubes with Rim

Manufactured from PP (Polypropylene) or PS (Polystyrene) for good chemical resistance. Kartell™ Disposable Centrifuge Tubes with Rim come with a round or conical bottom and are the ideal solution for centrifuge needs.

Brand:  Kartell™ 8831700

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 10555431

  • 229.00 € / Pack of 4000
Estimated Shipment: 27-08-2024
to see stock.
Add to basket

Description

Description

  • Disposable
  • Available in round or conical bottom
  • With rim
  • Available in PP or PS
  • For use with centrifuge
Specifications

Specifications

CE/IVD
5 mL
Laboratory use
87 mm
1300 RCF
Neutral
Polysulfone
Cylindrical
Centrifuge Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers