Learn More
Disposable Inoculation Loop, 1μL
Product Code. 12890155
Change view
Click to view available options
Volume:
1 μL
Unit Size:
Pack of 1000
Product Code. Volume unitSize
12890155 1 μL Pack of 1000
1 options
Product Code. 12890155 Supplier Microspec Supplier No. C3
Only null left
Add to basket
Add to basket
Manufactured from polystyrene for strength and flexibility. Microspec™ Disposable Inoculation Loops are ideal for the cultivation of microbes on plates by transferring inoculum for streaking and to transfer microscopic organisms.
  • High impact, polystyrene, disposable inoculating loops
  • 200mm long to reduce the risk of contamination
  • Includes 50 grip seal bags of 20 ea.
Specifications
Disposable Single-use
Volume 1 μL
Length (Metric) 200 mm
Material Plastic
Sterility Non-sterile
Product Type Inoculating Loop
For Use With (Equipment) Laboratory and Research use
Quantity 1000 Plastic Culture Loops (50 bags of 20)
For Use With (Application) Laboratory and Research use