missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disposable Luer-Lock Two-Part Syringe
Safe application of medication is ensured by the high quality of these needles. Henke Sass Wolf Hsw™ Disposable Luer-Lock Two-Part Syringes are designed for optimized user handling to ensure patient safety.
Brand: Henke Sass Wolf Hsw 4050X00V0
This item is not returnable.
View return policy
Description
- Available from sizes 1 to 50mL (60mL)
- Smooth flow, tight, high transparency barrel
- No rubber, styrene or DEHP
- Latex- and silicone-oil-free
- Pyrogen-free, PVC-free, non-toxic
- Sterile, individually blister strip packed
- Your first choice for an inert syringe
- Defined position of the plunger at volume “0” to feel when plunger is completely inserted
- LDS: no dead space for 1mL syringe
- Extended graduation for wider range of application
- Positive safety stop to prevent accidental spills
- 1 to 20mL (24mL) available also with by-packed needle
- 2 to 20mL available as Oral syringe with purple plunger
- 2mL to 50 mL (60mL) also available with Luer Lock
- 30mL and 50mL also available with Catheter tip
- All sizes available in non-sterile version (bulk) upon request
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction