missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Protective Coverall 4515, Orange
Description
- Type 5/6 Coverall designed to offer cost effective protection against hazardous dusts such as asbestos fibre
- Made from highly breathable SMS non-woven for effective release of body heat and moisture and reduced heat stress
- Soft and strong 50 gsm SMS for strong performance
- 2 panel hood, 2-way zip and storm flap with elasticated back and ankles to reduce the risk of skin exposure
- CE Cat III Type 5/6 hazardous dusts and limited liquid splashes
- EN 1073-2 nuclear particulate hold out
Specifications
Specifications
| Product Type | Protective Coveralls |
| Size | Large |
| Gender | Unisex |
| Material | SMS Polypropylene |
| Color | Orange |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Hood Type | 2-panel |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?