missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ Disposable PD-10 Desalting Columns

Product Code. 11768488 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric) Bed:
8.3 mL
Unit Size:
Pack of 30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Bed Unit Size
11768488 8.3 mL Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11768488 Supplier Cytiva™ Supplier No. 17085101
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use for desalting and buffer exchange, as well as removal of low-molecular weight compounds. Cytiva Life Sciences™ Disposable PD-10 Desalting Columns provide fast, high-recovery.

  • Packed with Sephadex™ G-25 medium
  • Designed for ease of use and equilibration
  • Prepare samples in range of 1.0 to 2.5mL by gravity flow or 1.75 to 2.5mL by centrifugation
  • Desalt multiple samples in parallel with desalting capacity >90%
  • Sample recovery in range of 70 to >95%
  • Includes adapters for centrifugation
TRUSTED_SUSTAINABILITY

Especificaciones

Product Type Desalting Column, Sephadex G-25 Medium Packing
Particle Size >50 μm (dry), 38 to 235 μm (wet)
Diameter (English) Inner Column 0.59 in.
For Use With (Application) Designed for ease of use and equilibration
Volume (Metric) Bed 8.3 mL
Content And Storage 4°C to 30°C
Diameter (Metric) Inner Column 1.5 cm
Quantity 30 Pk.
compliance-icons
Nombre del producto
  • Prepacked Disposable PD-10 Columns, 30 pcs
Palabra de advertencia
  • Warning
Clasificación
  • Skin sensitiser Category 1
Indicaciones de peligro
  • H317-May cause an allergic skin reaction.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Otros peligros
  • MIXTURE LIST-Contain: 5-chloro-2-methyl-4-isothiazolin-3-one,2-methyl-2H-isothiazol-3-one
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.