missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable PES Filter Units
GREENER_CHOICE

Product Code. 15993297 Shop All Fisherbrand Products
Change view
Click to view available options
Pore Size:
0.20 μm
0.45 μm
Volume (Metric):
150 mL
250 mL
500 mL
1000 mL
Diameter (Metric) Membrane:
50 mm
75 mm
90 mm
For Use With (Application):
Fluid clarification and particle removal
Sterile Filtration of media and serum
Unit Size:
Pack of 12
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Volume (Metric) Diameter (Metric) Membrane For Use With (Application) unitSize
15993297 0.20 μm 250 mL 50 mm Sterile Filtration of media and serum Pack of 12
15973297 0.20 μm 150 mL 50 mm Sterile Filtration of media and serum Pack of 12
15983297 0.45 μm 150 mL 50 mm Fluid clarification and particle removal Pack of 12
15903307 0.45 μm 250 mL 50 mm Fluid clarification and particle removal Pack of 12
15913307 0.20 μm 500 mL 75 mm Sterile Filtration of media and serum Pack of 12
15923307 0.45 μm 500 mL 75 mm Fluid clarification and particle removal Pack of 12
15933307 0.20 μm 1000 mL 90 mm Sterile Filtration of media and serum Pack of 12
15943307 0.45 μm 1000 mL 90 mm Fluid clarification and particle removal Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15993297
Only null left
Add to basket
Edge
Add to basket

Includes

Individually packaged sterile screw top style bottle cap

Increase cell culture sample protection with Fisherbrand sterile disposable vacuum filter units with PES membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.

  • PES membrane is low protein binding to maintain protein balance
  • PES membrane has low extractables to maintain media purity
  • Upper chamber of filter units is clearly marked with pore size, membrane type, catalog number, lot number and expiration date 
  • Non-pyrogenic and non-cytotoxic
  • Gamma radiation-sterilized and individually bagged for a 5-year sterile shelf life
  • 0.2μm units for sterile filtration
  • 0.45μm units for fluid clarification and particle removal

Certifications

Non-pyrogenic per USP Standard Lot-to-Lotnon-cytotoxic Lot-to-Lot sterility

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity Low binding
Certifications/Compliance Non-pyrogenic per USP Standard Lot-to-Lot
Disposable Single-use
Material (Membrane) PES
Pore Size 0.20 μm
Sterility Sterile
For Use With (Application) Sterile Filtration of media and serum
Type Filter Unit
Volume (Metric) 250 mL
Volume (Metric) Receiver 250 mL
Volume (Metric) Upper 250 mL
Diameter (Metric) Membrane 50 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.