missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Performance Plastics Disposable PVDF Flow Tubes

Product Code. 30284838
Click to view available options
Unit Size:
1 pack
Waterflow Rate:
0.03 to 2 L/min.
This item is not returnable. View return policy

Product Code. 30284838

Brand: Saint-Gobain Performance Plastics 0085PH000CX

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provide precise and reliable fluid measurement for biopharma and medical applications. Featuring PVDF and ruby wetted parts, these single-use tubes ensure gamma stability and resistance to corrosive substances. Ideal for critical processes.

  • Equflow Disposable PVDF Flow Tubes ensure precise, reliable fluid measurement for single-use applications.
  • Designed for biopharmaceutical and medical industries where accuracy and gamma stability are essential.
  • Constructed with PVDF and ruby wetted parts, providing exceptional durability and resistance to corrosion.
  • Highly suited for challenging environments, thanks to superior chemical resistance.
  • Supports gamma irradiation, enabling effective sterilization for sensitive applications.
  • Compatible with multiple mounting options like Tubeholder, Clamp and Clip Mounting for enhanced versatility.
  • Supplied in packs of ten, offering both convenience and cost-efficiency.
  • Optimized for critical processes requiring reliable performance and minimal contamination risk.
  • Sold separately from housing and sensor electronics, allowing tailored system configurations.
TRUSTED_SUSTAINABILITY

Specifications

Voltage 5 VDC
For Use With (Equipment) Flowmeter
Type Flow Tube
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.