missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Performance Plastics Disposable PVDF Flow Tubes

Product Code. 30284844
Change view
Click to view available options
Includes:
7 mm HB Clamp
Unit Size:
1 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
30284844 7 mm HB Clamp 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30284844 Supplier Saint-Gobain Performance Plastics Supplier No. 0045PH000CX

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Ruby and PVDF wetted parts for durability and chemical resistance, designed for various mounting models for single-use biopharma and medical applications

  • Single-use design for sterile and disposable applications
  • Made from PVDF with ruby wetted parts for durability
  • High chemical and mechanical resistance
  • Gamma sterilizable up to 50kGy
  • Compatible with multiple mounting styles
  • Sold in packs of 10
  • Housing and electronics sold separately
  • Suitable for biopharma and medical environments
TRUSTED_SUSTAINABILITY

Spécification

For Use With (Equipment) Tubeholder, Clamp System, Clip Mounting
Type Flow Tube
Includes 7 mm HB Clamp
Airflow Rate 0.03 to 2 L/min.
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.