Learn More
Saint-Gobain Performance Plastics Disposable PVDF Flow Tubes
Product Code. 30284851
Change view
Click to view available options
Includes:
1/8 NPT Single Use Tubes
Unit Size:
1 pack
Product Code. Includes Unit Size
30284851 1/8 NPT Single Use Tubes 1 pack
1 options
Product Code. 30284851 Supplier Saint-Gobain Performance Plastics Supplier No. 0045PN000TX
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Ruby and PVDF wetted parts for durability and chemical resistance, designed for various mounting models for single-use biopharma and medical applications
  • Single-use design for sterile and disposable applications
  • Made from PVDF with ruby wetted parts for durability
  • High chemical and mechanical resistance
  • Gamma sterilizable up to 50kGy
  • Compatible with multiple mounting styles
  • Sold in packs of 10
  • Housing and electronics sold separately
  • Suitable for biopharma and medical environments
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Tubeholder, Clamp System, Clip Mounting
Type Flow Tube
Includes 1/8 NPT Single Use Tubes
Airflow Rate 0.03 to 2 L/min.