Learn More
New Source™ Disposable Chemical Resistant SMS Coveralls Type 5/6
Click to view available options
Size:
2X-Large
3X-Large
Large
Medium
X-Large
Unit Size:
Pack of 50
Description
- Conforms to CE Category III Type 5/6
- Non-woven SMS disposable fabric
- Protects against chemical and biological hazards
Specifications
Specifications
| Size | Medium |
| Material | Non-woven SMS |
| Disposable | Single-use |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Closure Type | Flap |
| Certifications/Compliance | CE Cat III PPE meeting: EN ISO 13982-1:2004, EN ISO 13034: 2005 +A1:2009, EN ISO 14325:2004 |
| Weight (Metric) | 55 gsm |