Learn More
Heinz Herenz Dissection Sets
Product Code. 30158371
Change view
Click to view available options
Disposable:
Reusable
Unit Size:
Each
Product Code. Disposable Unit Size
30158371 Reusable Each
1 options
Product Code. 30158371 Supplier Heinz Herenz Supplier No. 1110401
  • ONLINE EXCLUSIVE PRICE
    49.99 €
    30.90 € / Pack of 1
    Save 19.09 €
    38% Off
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Dissection sets with wooden case.
  • Dissection sets
  • Strong wooden case: 19 x 9 x 3.5 cm
  • Contents: 1 scalpel handle No. 4 with 2 blades, 1 scalpel, 1 scalpel handle No. 3 with 2 blades, 1 forceps, 1 scissors (pointed), 2 dissection needles (1x pointed, 1x with blade)
Specifications
Product Type Dissection Set
Disposable Reusable
Sterility Non-sterile