missing translation for 'onlineSavingsMsg'
Learn More

Tristel™ Distel™ Surface Disinfectant

Product Code. 12859357
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
5000 mL
Composition:
Distel TM301
Distel TM305
Distel TM309
Container Type:
Spray bottle
Unit Size:
5000mL
500mL
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Composition Container Type Unit Size
12859357 500 mL Distel TM301 Spray bottle 500mL
12879357 1000 mL Distel TM305 - Pack of 1000
12899357 5000 mL Distel TM309 - 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12859357 Supplier Tristel™ Supplier No. TM301

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tristel™ Distel™ Surface Disinfectant is an all-purpose, non-alcohol based formula for use in cleaning and disinfecting laboratory surfaces and equipment.

  • Classified as a biocide under the EU Biocides Regulation
  • Tested and approved as bactericidal, fungicidal, virucidal and tuberculocidal
  • Effective in the presence of blood borne diseases (HIV/Hepatitis B) and also destroys DNA and RNA
  • Recommended for use at a dilution of 1:100 for all normal laboratory requirements
  • Suitable for use on most surfaces including glass, metal, plastic, rubber and fabric

Specifications

Product Type Surface Disinfectant
Composition Distel TM301
Container Type Spray bottle
For Use With (Application) For professional use only
Volume (Metric) 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.