missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tristel™ Distel™ Surface Disinfectant
Description
- Classified as a biocide under the EU Biocides Regulation
- Tested and approved as bactericidal, fungicidal, virucidal and tuberculocidal
- Effective in the presence of blood borne diseases (HIV/Hepatitis B) and also destroys DNA and RNA
- Recommended for use at a dilution of 1:100 for all normal laboratory requirements
- Suitable for use on most surfaces including glass, metal, plastic, rubber and fabric
Specifications
Specifications
| Product Type | Surface Disinfectant |
| Form | Concentrate |
| Composition | Distel TM305 |
| Dilution | 0.11111111111 |
| For Use With (Application) | For professional use only |
| Volume (Metric) | 1000 mL |
Safety and Handling
Product Identifier
- DISTEL HIGH LEVEL LABORATORY DISINFECTANT
- Danger
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1B
- Serious eye damage/eye irritation Category 1
- Carcinogenicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?