missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Distilled Water
Suitable for use in solvents and in analytical chemistry.
Brand: N/A W14156
This item is not returnable.
View return policy
Description
- Molecular Formula: H2O
- Molar Mass: 18.02g/mol
- CAS: 7732-18-5
- EIN ECS No.: 231-791-2
Specifications
| 7732-18-5 | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| O | |
| 18.015 | |
| CHEBI:15377 | |
| 23hPa (at 20°C) |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| oxidane | |
| 962 | |
| 18.02g/mol | |
| 1.00g/cm3 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction