missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Distilling Links with Liebig Condenser and Head to Claisen

Distilling Links with Liebig Condenser and Head to Claisen made of DURAN™ tubing.

Brand:  Lenz Laborglasinstrumente™ 05057201

Additional Details : Weight : 0.60000kg

 View more versions of this product

Product Code. 10040511

  • 84.80 € / Each
Estimated Shipment: 25-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

For distillation thermometers of 55 mm built-In length
Duran Glass
160 mm
Distilling Links
NS 14/23, NS 14/23
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers