Learn More
Lenz Laborglasintrumente Distilling Receivers Acc. to Anschütz-Thiele, Bent
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
309.31 € - 348.85 €
Specifications
Includes | Bent adapter, graduated, with NS socket and NS cone, vacuum connection: thread GL18 |
---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Material | Price | Quantity & Availability | |||
10579041
|
Lenz Laborglasintrumente
05298004 |
25 mL | Socket and Cone NS 29/32 | Duran Glass |
329.02 €
Each |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10030231
|
Lenz Laborglasintrumente
05298101 |
25 mL | Socket and Cone NS 14/23 | Duran PTFE |
320.65 €
Each |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10431093
|
Lenz Laborglasintrumente
05298014 |
100 mL | Socket and Cone NS 29/32 | Duran Glass |
338.35 €
Each |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10623242
|
Lenz Laborglasintrumente
05298001 |
25 mL | Socket and Cone NS 14/23 | Duran Glass |
309.31 €
Each |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10673622
|
Lenz Laborglasintrumente
05298114 |
100 mL | Socket and Cone NS 29/32 | Duran PTFE |
348.85 €
Each |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10768461
|
Lenz Laborglasintrumente
05298104 |
50 mL | Socket and Cone NS 29/32 | Duran PTFE |
340.37 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Receivers with bent adapter, graduated
- With NS socket and NS cone
- vacuum connection: thread GL18
- NS stopcock with solid glass or PTFE plug
- Made of DURAN™ tubing
Specifications
Bent adapter, graduated, with NS socket and NS cone, vacuum connection: thread GL18 |