Learn More
Lenz Laborglasintrumente Distilling Receivers Acc.to Bernauer with Flasks
Product Code. 10306361
Change view
Click to view available options
Standard Taper Joint:
Upper Socket 12/23 NS
Upper Socket 19/26 NS
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
10306361 Upper Socket 12/23 NS Each
10326361 Upper Socket 19/26 NS Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10306361 Supplier Lenz Laborglasintrumente Supplier No. 05288001
  • ONLINE EXCLUSIVE PRICE
    157.00 €
    102.99 € / Pack of 1
    Save 54.01 €
    34% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Distilling receivers acc. to Bernauer (modified)
  • With three cones NS 14/23 and three flasks
  • Vacuum connection with glass olive
  • Made of DURAN™ tubing
Specifications
Material Duran Glass
Capacity (Metric) 50 mL
Quantity 1 Ea.
Standard Taper Joint Upper Socket 12/23 NS