missing translation for 'onlineSavingsMsg'
Learn More

Disuccinimidyl suberate, 97%

Product Code. 15498217
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498217 500 mg 500mg
15488217 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498217 Supplier Thermo Scientific Alfa Aesar Supplier No. H51771.ME
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68528-80-3 | C16H20N2O8 | 368.342 g/mol

Disuccinimidyl suberate is a homobifunctional amine-reactive crosslinker. It is usually coupled to molecules containing primary amines via amide bonds buffered at pH 7.5 (6.5-8.5). It incorporates an 8-atom spacer arm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Disuccinimidyl suberate is a homobifunctional amine-reactive crosslinker. It is usually coupled to molecules containing primary amines via amide bonds buffered at pH 7.5 (6.5-8.5). It incorporates an 8-atom spacer arm.

Solubility
Insoluble in water. Soluble in chloroform: 50 mg/mL.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68528-80-3
Molecular Formula C16H20N2O8
Molecular Weight (g/mol) 368.342
MDL Number MFCD00049059
InChI Key ZWIBGKZDAWNIFC-UHFFFAOYSA-N
Synonym disuccinimidyl suberate, bis 2,5-dioxopyrrolidin-1-yl octanedioate, suberic acid bis n-hydroxysuccinimide ester, di n-succinimidyl suberate, bicl100, 2,5-pyrrolidinedione, 1,1'-1,8-dioxo-1,8-octanediyl bis oxy bis, nhs-sa, disuccinimidylsuberate, dsis, n-hydroxysuccinimide suberic acid ester
PubChem CID 100658
IUPAC Name bis(2,5-dioxopyrrolidin-1-yl) octanedioate
SMILES C1CC(=O)N(C1=O)OC(=O)CCCCCCC(=O)ON2C(=O)CCC2=O

Specifications

Melting Point 166°C to 170°C
Quantity 500 mg
Solubility Information Insoluble in water. Soluble in chloroform: 50mg/mL.
Formula Weight 368.34
Percent Purity 97%
Chemical Name or Material Disuccinimidyl suberate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.