missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DTE, Dithioerythritol, Electrophoresis, Fisher BioReagents™
Beskrivelse
- Used as reducing agent
- Reduces disulfide bonds in proteins and peptides
- Functionally tested for use in electrophoresis
- Added in low concentrations, DTE used as an enzyme stabilizer during protein purification
- Hydroxyl (-OH) groups of DTE are in the cisform, as opposed to transfor DTT
Tekniske data
Tekniske data
| CAS | 6892-68-8 |
| Absorbance | 0.13 AU max. |
| Melting Point | 82°C |
| Color | White |
| Assay Percent Range | ≥99 % |
| Packaging | Poly Bottle |
| Molecular Formula | C4H10O2S2 |
| Linear Formula | HSCH2CH(OH)CH(OH)CH2SH |
| MDL Number | MFCD00063750 |
| Quantity | 5 g |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- Dithioerythritol
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?