missing translation for 'onlineSavingsMsg'
Learn More

Dithiooxamide, 98%

Product Code. 11466666
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466666 5 g 5g
11476666 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466666 Supplier Thermo Scientific Alfa Aesar Supplier No. B24866.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79-40-3 | C2H4N2S2 | 120.188 g/mol

Dithiooxamide acts as a chelating agent and used in the determination of copper(II), nickel(II) and cobalt(II). It is used as a building block in the synthesis of cyclen. It is involved in the preparation of thiazolothiazole-linked porous organic polymers and N,N'-disubstituted dithiooxamides. It is also employed as a modifier to prepare the modified glassy carbon electrode which is used to investigate the electrochemical properties of quercetin, an important flavonoid derivative. Further, it is involved in the preparation of chelating resin with formaldehyde, which finds application in separation and concentration of silver ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dithiooxamide acts as a chelating agent and used in the determination of copper(II), nickel(II) and cobalt(II). It is used as a building block in the synthesis of cyclen. It is involved in the preparation of thiazolothiazole-linked porous organic polymers and N,N′-disubstituted dithiooxamides. It is also employed as a modifier to prepare the modified glassy carbon electrode which is used to investigate the electrochemical properties of quercetin, an important flavonoid derivative. Further, it is involved in the preparation of chelating resin with formaldehyde, which finds application in separation and concentration of silver ions.

Solubility
Soluble in acetone and chloroform. Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-40-3
Molecular Formula C2H4N2S2
Molecular Weight (g/mol) 120.188
MDL Number MFCD00004941
InChI Key OAEGRYMCJYIXQT-UHFFFAOYSA-N
Synonym dithiooxamide, rubeanic acid, ethanebis thioamide, oxamide, dithio, dithioxamide, rubeane, rubean, hydrorubeanic acid, oxaldiimidic acid, dithio, usaf mk-6
PubChem CID 2777982
IUPAC Name ethanedithioamide
SMILES C(=S)(C(=S)N)N

Specifications

Melting Point 245°C (decomposition)
Quantity 5 g
Solubility Information Soluble in acetone and chloroform. Insoluble in water.
Formula Weight 120.2
Percent Purity 98%
Chemical Name or Material Dithiooxamide
compliance-icons
Product Identifier
  • Dithiooxamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Ethanedithioamide

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.