missing translation for 'onlineSavingsMsg'
Learn More

Divinyl Sulfone (Stabilized with Hydroquinone), TRC

Produktkod. 30381687
Change view
Klicka för att se tillgängliga alternativ
Quantity:
100 mg
Förpackningsstorlek:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
30381687 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. View return policy
Product Code. 30381687 Supplier Toronto Research Chemicals Supplier No. TRCD679243100MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Divinyl Sulfone
CAS 77-77-0
Molecular Formula C4H6O2S
Quantity 100 mg
Synonym DivinylSulfone
InChI Formula InChI=1S/C4H6O2S/c1-3-7(5,6)4-2/h3-4H,1-2H2
SMILES C=CS(=O)(=O)C=C
IUPAC Name 1-ethenylsulfonylethene
Molecular Weight (g/mol) 118.16
Formula Weight 118.0089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.