missing translation for 'onlineSavingsMsg'
Learn More

Divinylbenzene (ortho-, meta-, or para- substituted) (~80%), TRC

Product Code. 30368991
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368991 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368991 Supplier Toronto Research Chemicals Supplier No. TRCD49487310ML
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Divinylbenzene (ortho-, meta-, or para- substituted)
CAS 1321-74-0
Molecular Formula C10H10
Quantity 10 mL
Synonym Divinylbenzene (mixture of isomers), Benzene, diethenyl-, Benzene, divinyl- (8CI), Diethenylbenzene, DVB 96, DVB 960, DVB HP, DVB-H, DVR 960, Divinylbenzene, Vinylstyrene
InChI Formula InChI=1S/C10H10/c1-3-9-7-5-6-8-10(9)4-2/h3-8H,1-2H2
SMILES C=CC1=CC=C(C=C)C=C1.C=CC1=CC=CC=C1C=C
Molecular Weight (g/mol) 130.19
Formula Weight 390.2348
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.