missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ Divosan Activ Disinfectant

Product Code. 15320308
Change view
Click to view available options
Volume (Metric):
20 L
Unit Size:
22.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15320308 20 L 22.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15320308 Supplier Diversey™ Supplier No. 6028738
  • ONLINE EXCLUSIVE PRICE
    166.00 €
    135.50 € / 22.5 L
    Save 30.50 €
    18% Off
View all promotions
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Disinfects with a 5% peracetic acid composition. Diversey Divosan Activ Disinfectant is a stabilized parectic acid solution that that non-foaming and completely free-wrinsing. It is effective against all types of micro-organisms.

  • Effective against all types of micro-organisms including bacteria, yeasts, fungi, spores, and viruses
  • Use at concentrations between 0.1-4% w/w depending upon application
  • Rinse thoroughly after use

Specifications

pH <2
Sterility Non-sterile
Composition 5% paracetic acid
Dilution 0.1-4% w/w
Filtered Unfiltered
Volume (Metric) 20 L
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Divosan Activ VT5
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: hydrogen peroxide, acetic acid, peracetic acid
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.