missing translation for 'onlineSavingsMsg'
Learn More

Dizocilpine maleate, MedChemExpress

Product Code. 16476308
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476308 10 mg 10mg
16452427 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16476308 Supplier Medchem Express Supplier No. HY15084
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dizocilpine maleate (MK-801 maleate) is a potent, selective and non-competitive NMDA receptor antagonist with Kd of 37.2 nM in rat brain membranes.

Dizocilpine maleate (MK-801 maleate) is a potent, selective and non-competitive NMDA receptor antagonist with Kd of 37.2 nM in rat brain membranes.

Specifications

Chemical Name or Material Dizocilpine maleate
CAS 77086-22-7
Color White
Purity Grade Notes Research
Molecular Formula C20H19NO4
Quantity 10 mg
Synonym MK-801 maleate
Solubility Information DMSO : 133.33 mg/mL (395.20 mM; Need ultrasonic) ∣Ethanol : 25 mg/mL (74.10 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@]12C3=CC=CC=C3C[C@@H](N2)C4=CC=CC=C14.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 337.37
Formula Weight 337.37
Percent Purity 98.39%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.